Anti-CRD1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66869-100UL A66869-50UL
77835-776EA 521.4 CAD
77835-776 77835-777
Anti-CRD1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CRD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR