Anti-Trypsin inhibitor DE-3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A51671-100UG A51671-50UG
77817-614EA 521.4 CAD
77817-614 77817-615
Anti-Trypsin inhibitor DE-3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Trypsin inhibitor DE-3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Erythrina caffra
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR