Anti-thiO Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A52572-100UG A52572-50UG
77819-209EA 521.4 CAD
77819-209 77819-210
Anti-thiO Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: thiO
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus subtilis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR