Anti-SMUG1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A57187-100UG A57187-50UG
77827-655EA 521.4 CAD
77827-655 77827-656
Anti-SMUG1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: SMUG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol FDG;HMUDG;MGC104370;Single strand selective monofunctional uracil DNA glycosylase 1;Single strand selective monofunctional uracil DNA glycosylase;Single-strand selective monofunctional uracil DNA glycosylase;SMUG 1;Smug1;SMUG1 protein;SMUG1_HUMAN;UNG 3;UNG3
Environmentally Preferable
Antigen name Single-strand selective monofunctional uracil DNA glycosylase protein (1-177AA)
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q53HV7
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR