Anti-PDE6D Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A52664-50UG A52664-100UG
77819-386EA 355.5 CAD
77819-386 77819-385
Anti-PDE6D Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PDE6D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR