Anti-SLC39A8 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A35038-100UG A35038-50UG
77800-076EA 521.4 CAD
77800-076 77800-077
Anti-SLC39A8 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: SLC39A8
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol SLC39A8
Antigen name Zinc transporter ZIP8 protein (30-132AA)
Environmentally Preferable
Antigen synonyms SLC39A8|BIGM103|ZIP8|PP3105|Zinc transporter ZIP8|BCG-induced integral membrane protein in monocyte clone 103 protein|LIV-1 subfamily of ZIP zinc transporter 6|LZT-Hs6|Solute carrier family 39 member 8|Zrt- and Irt-like protein 8|ZIP-8
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9C0K1
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR