Anti-RHCG Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A69209-100UG A69209-50UG
77840-169EA 521.4 CAD
77840-169 77840-170
Anti-RHCG Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: RHCG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR