Anti-FAM76B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A57092-100UG A57092-50UG
77827-476EA 521.4 CAD
77827-476 77827-477
Anti-FAM76B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: FAM76B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR