Anti-CNGB1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A55907-100UG A55907-50UG
77825-272EA 521.4 CAD
77825-272 77825-273
Anti-CNGB1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CNGB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR