Anti-PDE9A Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A52709-100UG A52709-50UG
77819-469EA 521.4 CAD
77819-469 77819-470
Anti-PDE9A Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PDE9A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR