Anti-PPP1CC Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A31811-100UG A31811-50UG
77795-072EA 521.4 CAD
77795-072 77795-073
Anti-PPP1CC Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:2000 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: PPP1CC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR