Anti-NPY Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43392-100UL A43392-50UL
77812-974EA 521.4 CAD
77812-974 77812-975
Anti-NPY Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000

Type:
Antigen: NPY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR