Anti-cpg2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51613-100UG A51613-50UG
77817-502EA 521.4 CAD
77817-502 77817-503
Anti-cpg2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: cpg2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pseudomonas sp.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR