Anti-TNKS1BP1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A68163-100UG A68163-50UG
77838-203EA 521.4 CAD
77838-203 77838-204
Anti-TNKS1BP1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: TNKS1BP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR