Anti-COX19 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A56625-100UG A56625-50UG
77826-623EA 521.4 CAD
77826-623 77826-624
Anti-COX19 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: COX19
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR