Anti-SLURP1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A35144-100UG A35144-50UG
77800-244EA 521.4 CAD
77800-244 77800-245
Anti-SLURP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLURP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR