Anti-RNF103 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A33311-100UG A33311-50UG
77797-447EA 521.4 CAD
77797-447 77797-448
Anti-RNF103 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200

Type:
Antigen: RNF103
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR