Anti-PIM3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A59609-100UG A59609-50UG
77832-215EA 521.4 CAD
77832-215 77832-216
Anti-PIM3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PIM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR