Anti-PDE6D Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A52665-100UG A52665-50UG
77819-387EA 521.4 CAD
77819-387 77819-388
Anti-PDE6D Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PDE6D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR