Anti-TLR5 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A36683-100UG A36683-50UG
77802-701EA 521.4 CAD
77802-701 77802-702
Anti-TLR5 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TLR5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR