Anti-SLC13A2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A54919-100UL A54919-50UL
77823-396EA 521.4 CAD
77823-396 77823-397
Anti-SLC13A2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: SLC13A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR