Anti-PRPS2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A32137-100UG A32137-50UG
77795-598EA 521.4 CAD
77795-598 77795-599
Anti-PRPS2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PRPS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR