Anti-TNF Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A43376-50UG A43376-100UG
77812-950EA 355.5 CAD
77812-950 77812-949
Anti-TNF Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TNF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR