Anti-THRB Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A36567-100UG A36567-50UG
77802-520EA 521.4 CAD
77802-520 77802-521
Anti-THRB Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: THRB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR