Anti-MS4A12 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A70437-50UL A70437-100UL
77842-498EA 355.5 CAD
77842-498 77842-497
Anti-MS4A12 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: MS4A12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR