Anti-FBXW10 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A57711-100UG A57711-50UG
77828-637EA 521.4 CAD
77828-637 77828-638
Anti-FBXW10 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: FBXW10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol FBXW10
Antigen name F-box/WD repeat-containing protein 10 protein (701-1000AA)
Environmentally Preferable
Antigen synonyms FBXW10| F-box/WD repeat-containing protein 10| F-box and WD-40 domain-containing protein 10| Ubiquitin ligase-specificity factor
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q5XX13
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR