Anti-thiO Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A52570-50UG A52570-100UG
77819-206EA 355.5 CAD
77819-206 77819-205
Anti-thiO Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: thiO
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus subtilis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR