Anti-LHPP Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A67543-100UG A67543-50UG
77837-041EA 521.4 CAD
77837-041 77837-042
Anti-LHPP Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: LHPP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR