Anti-RHOXF2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66556-100UG A66556-50UG
77835-180EA 521.4 CAD
77835-180 77835-181
Anti-RHOXF2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: RHOXF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR