Anti-CRP Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A41727-100UG A41727-50UG
77809-441EA 521.4 CAD
77809-441 77809-442
Anti-CRP Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CRP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR