Anti-SDHA Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A33397-100UG A33397-50UG
77797-594EA 521.4 CAD
77797-594 77797-595
Anti-SDHA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SDHA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR