Anti-HINT1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A41227-100UG A41227-50UG
77808-786EA 521.4 CAD
77808-786 77808-787
Anti-HINT1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: HINT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR