Anti-thiO Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A52569-100UG A52569-50UG
77819-203EA 521.4 CAD
77819-203 77819-204
Anti-thiO Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: thiO
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus subtilis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR