Anti-CRD1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A66871-100UL A66871-50UL
77835-780EA 521.4 CAD
77835-780 77835-781
Anti-CRD1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CRD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
New Product
Order Now

SPECIFICATIONS

Antigen symbol CRD1
Antigen name Recombinant Arabidopsis thaliana Magnesium-protoporphyrin IX monomethyl ester [oxidative] cyclase, chloroplastic protein (37-409AA)
Environmentally Preferable
Antigen synonyms CRD1|ACSF|AT103|CHL27|ZIP|At3g56940|F24I3.20Magnesium-protoporphyrin IX monomethyl ester [oxidative] cyclase|chloroplastic|Mg-protoporphyrin IX monomethyl ester oxidative cyclase|EC 1.14.13.81|Copper response defect 1 protein|Dicarboxylate diiron protein|AtZIP|MPE-cyclase
Clonality Recombinant Polyclonal
Conjugation Biotin
Reactivity A. thaliana
Host Rabbit
Gene ID Q9M591
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR