Anti-COX10 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A55373-100UL A55373-50UL
77824-252EA 521.4 CAD
77824-252 77824-253
Anti-COX10 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: COX10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR