Anti-KCND2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A68574-100UL A68574-50UL
77838-978EA 521.4 CAD
77838-978 77838-979
Anti-KCND2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: KCND2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR