Anti-SLC25A6 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A34952-100UG A34952-50UG
77799-931EA 521.4 CAD
77799-931 77799-932
Anti-SLC25A6 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC25A6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR