Anti-DCXR Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A59649-100UL A59649-50UL
77832-292EA 521.4 CAD
77832-292 77832-293
Anti-DCXR Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: DCXR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR