Anti-ERBIN Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66112-100UL A66112-50UL
77834-354EA 521.4 CAD
77834-354 77834-355
Anti-ERBIN Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: ERBIN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR