Anti-Beta-mammal toxin Css4 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A51183-100UG A51183-50UG
77816-693EA 521.4 CAD
77816-693 77816-694
Anti-Beta-mammal toxin Css4 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Beta-mammal toxin Css4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Centruroides suffusus suffusus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR