Anti-RASA2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A55735-100UL A55735-50UL
77824-946EA 521.4 CAD
77824-946 77824-947
Anti-RASA2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: RASA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR