Anti-GJB7 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A58900-100UG A58900-50UG
77830-888EA 521.4 CAD
77830-888 77830-889
Anti-GJB7 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: GJB7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR