Anti-OGFOD2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A58740-100UG A58740-50UG
77830-584EA 521.4 CAD
77830-584 77830-585
Anti-OGFOD2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: OGFOD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR