Anti-Major pepsin inhibitor 3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50482-100UG A50482-50UG
77815-375EA 521.4 CAD
77815-375 77815-376
Anti-Major pepsin inhibitor 3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Major pepsin inhibitor 3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Ascaris suum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR