Anti-ASNSD1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A70422-100UG A70422-50UG
77842-470EA 521.4 CAD
77842-470 77842-471
Anti-ASNSD1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ASNSD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR