Anti-Beta-lactamase inhibitory protein Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50685-100UG A50685-50UG
77815-763EA 521.4 CAD
77815-763 77815-764
Anti-Beta-lactamase inhibitory protein Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Beta-lactamase inhibitory protein
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptomyces clavuligerus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR