Anti-STRADB Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A68730-100UG A68730-50UG
77839-274EA 521.4 CAD
77839-274 77839-275
Anti-STRADB Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: STRADB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR