Anti-PSMG3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66580-100UG A66580-50UG
77835-225EA 521.4 CAD
77835-225 77835-226
Anti-PSMG3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: PSMG3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR