Anti-HERC5 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A69933-100UG A69933-50UG
77841-541EA 521.4 CAD
77841-541 77841-542
Anti-HERC5 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: HERC5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR