Anti-CELSR1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A67840-100UL A67840-50UL
77837-608EA 521.4 CAD
77837-608 77837-609
Anti-CELSR1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CELSR1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR